Learn More
AdipoGen Aphidicolin 17-acetate
SDP
Catalog No. 502561985
Encompass_Preferred
Chemical. CAS 51103-57-2. Formula C22H36O5. MW 380.5. Isolated from Phoma sp. BS 7210. Phytotoxin inhibits root growth of seeding. Specific inhibitor of eukaryotic DNA polymerase alpha and DNA synthesis competition with dCTP.
Catalog No. 50-256-1985 Supplier AdipoGen Supplier No. BVT0337C250
Chemical. CAS 51103-57-2. Formula C22H36O5. MW 380.5. Isolated from Phoma sp. BS 7210. Phytotoxin inhibits root growth of seeding. Specific inhibitor of eukaryotic DNA polymerase alpha and DNA synthesis competition with dCTP.