Learn More
TARGETMOL CHEMICALS INC Aphidicolin 1MG
SDP
Catalog No. 502879568
Encompass_Preferred
Aphidicolin, an antibiotic derived from the mold Cephalosporium aphidicola, is a potent inhibitor of DNA (deoxyribonucleic acid) synthesis, effectively hindering the proliferation of herpes simplex virus[2]. It targets DNA polymerase alpha and delta, disrupting mitotic cell division by affecting DNA polymerase activity[1]. Additionally, aphidicolin enhances apoptosis when combined with arabinosyl nucleosides in a human promyelocytic leukemia cell line[3].
Catalog No. 50-287-9568 Supplier TARGETMOL CHEMICALS INC Supplier No. T143041MG
Aphidicolin, an antibiotic derived from the mold Cephalosporium aphidicola, is a potent inhibitor of DNA (deoxyribonucleic acid) synthesis, effectively hindering the proliferation of herpes simplex virus[2]. It targets DNA polymerase alpha and delta, disrupting mitotic cell division by affecting DNA polymerase activity[1]. Additionally, aphidicolin enhances apoptosis when combined with arabinosyl nucleosides in a human promyelocytic leukemia cell line[3].