Learn More
Sigma Aldrich Fine Chemicals Biosciences Aphidicolin - CAS 38966-21-1 - Calbiochem Aphidicolin, CAS 38966-21-1, is a cell-permeable antibiotic that acts as a cell synchronization agent. Blocks the cell cycle at early S-phase. | 38966-21-1 | MFCD00083214 | 1MG
Catalog No. NC1952274
Encompass
Aphidicolin - CAS 38966-21-1 - Calbiochem Aphidicolin, CAS 38966-21-1, is a cell-permeable antibiotic that acts as a cell synchronization agent. Blocks the cell cycle at early S-phase. | Purity: >=98% (TLC) | Mol Wt: 338.48 | 38966-21-1 | MFCD00083214 | 1MG
Catalog No. NC1952274 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 1782731MG
Aphidicolin - CAS 38966-21-1 - Calbiochem Aphidicolin, CAS 38966-21-1, is a cell-permeable antibiotic that acts as a cell synchronization agent. Blocks the cell cycle at early S-phase. | Purity: >=98% (TLC) | Mol Wt: 338.48 | 38966-21-1 | MFCD00083214 | 1MG