Learn More
AdipoGen -Aphidicolin
SDP
Catalog No. 502562000
Encompass_Preferred
Chemical. CAS 38966-21-1. Formula C20H34O4. MW 338.5. Isolated from Phoma sp. BS 7210. Phytotoxin. Antibiotic. Antiviral and antineoplastic agent. Antileishmanial agent. Reversible inhibitor of eukaryotic nuclear DNA replication. Useful for cell synchronization. Blocks the cell cycle at G1/S phase. Prolongs the half life of DNA methyltransferase. DNA methylation/demethylation modulator. Specific DNA polymerase alpha and delta inhibitor in eukaryotic cells and in some viruses of animal origin. Acts synergistically with vincristine and doxorubicin. Apoptosis inhibitor/inducer.
Catalog No. 50-256-2000 Supplier AdipoGen Supplier No. BVT0307M025
Add to Cart
Add to Cart
Chemical. CAS 38966-21-1. Formula C20H34O4. MW 338.5. Isolated from Phoma sp. BS 7210. Phytotoxin. Antibiotic. Antiviral and antineoplastic agent. Antileishmanial agent. Reversible inhibitor of eukaryotic nuclear DNA replication. Useful for cell synchronization. Blocks the cell cycle at G1/S phase. Prolongs the half life of DNA methyltransferase. DNA methylation/demethylation modulator. Specific DNA polymerase alpha and delta inhibitor in eukaryotic cells and in some viruses of animal origin. Acts synergistically with vincristine and doxorubicin. Apoptosis inhibitor/inducer.