Learn More
Medchemexpress LLC Aphidicolin | 38966-21-1 | 50 MG
SDP
Catalog No. 5000419453
Encompass_Preferred
Aphidicolin is an inhibitor of DNA polymerase α and δ, which prevents mitotic cell division by interfering with DNA polymerase activity. It is an antibiotic produced by the mold *Cephalosporium aphidicola* and inhibits cellular deoxyribonucleic acid synthesis and the growth of herpes simplex virus. Aphidicolin also exhibits anti-orthopoxvirus activity and potentiates apoptosis induced by arabinosyl nucleosides in a human promyelocytic leukemia cell line.
  • Inhibitor of DNA polymerase α and δ
  • Prevents mitotic cell division
  • Antibiotic produced by *Cephalosporium aphidicola*
  • Inhibits cellular deoxyribonucleic acid synthesis
  • Inhibits growth of herpes simplex virus
  • Exhibits anti-orthopoxvirus activity
  • Potentiates apoptosis induced by arabinosyl nucleosides
Catalog No. 50-004-19453 Supplier Medchemexpress LLC Supplier No. HYN673350MG
Aphidicolin is an inhibitor of DNA polymerase α and δ, which prevents mitotic cell division by interfering with DNA polymerase activity. It is an antibiotic produced by the mold *Cephalosporium aphidicola* and inhibits cellular deoxyribonucleic acid synthesis and the growth of herpes simplex virus. Aphidicolin also exhibits anti-orthopoxvirus activity and potentiates apoptosis induced by arabinosyl nucleosides in a human promyelocytic leukemia cell line.
  • Inhibitor of DNA polymerase α and δ
  • Prevents mitotic cell division
  • Antibiotic produced by *Cephalosporium aphidicola*
  • Inhibits cellular deoxyribonucleic acid synthesis
  • Inhibits growth of herpes simplex virus
  • Exhibits anti-orthopoxvirus activity
  • Potentiates apoptosis induced by arabinosyl nucleosides