Learn More
Cayman Chemical (+)-Aphidicolin, Purity: ≥98%, CAS: 38966-21-1, 5 mg
SDP
Catalog No. 502953983
Encompass_Preferred
A natural tetracyclic diterpene with antiviral activity against herpes simplex; a cell-permeable, reversible inhibitor of DNA replication in eukaryotic cells, specifically blocking the activity of DNA polymerases α, δ, and ε when used at low micromolar levels.
Catalog No. 50-295-3983 Supplier Cayman Chemical Supplier No. 140075 MG
A natural tetracyclic diterpene with antiviral activity against herpes simplex; a cell-permeable, reversible inhibitor of DNA replication in eukaryotic cells, specifically blocking the activity of DNA polymerases α, δ, and ε when used at low micromolar levels.