Learn More
Surmodics Inc Alkaline Phosphatase/Horseradish Peroxidase Block, 100mL
SDP
Catalog No. NC1391159
Encompass_Preferred
BioFX™ Alkaline Phosphatase/Horseradish Peroxidase Block is supplied as a ready-to-use blocking reagent for immunohistochemistry samples. The solution blocks both alkaline phosphatase and peroxidase endogenous enzymatic activity.
Catalog No. NC1391159 Supplier Surmodics Inc Supplier No. APHP010001
BioFX™ Alkaline Phosphatase/Horseradish Peroxidase Block is supplied as a ready-to-use blocking reagent for immunohistochemistry samples. The solution blocks both alkaline phosphatase and peroxidase endogenous enzymatic activity.