Learn More
Medchemexpress LLC API-1 | 36707-00-3 | MFCD18086904 | 99.6% | 337.29 g/mol | C13H15N5O6 | 5 MG
SDP
Catalog No. 5000263850
Encompass_Preferred
API-1 is a small-molecule, selective Akt/PKB inhibitor used in research to probe Akt signaling, apoptosis, and cell proliferation. It binds the pleckstrin homology (PH) domain to inhibit Akt membrane translocation, lowers phosphorylated Akt levels, and is supplied as a high-purity reagent for in vitro studies.
  • Selective Akt/PKB inhibition for pathway studies.
  • Binds the pleckstrin homology (PH) domain.
  • Reduces phosphorylated Akt and cell proliferation in assays.
  • High purity (≈99.6%) suitable for biochemical experiments.
  • Small package sizes available for screening and pilot studies.
  • Molecular weight 337.29 g/mol; formula C13H15N5O6.
Catalog No. 50-002-63850 Supplier Medchemexpress LLC Supplier No. HY1100775MG
API-1 is a small-molecule, selective Akt/PKB inhibitor used in research to probe Akt signaling, apoptosis, and cell proliferation. It binds the pleckstrin homology (PH) domain to inhibit Akt membrane translocation, lowers phosphorylated Akt levels, and is supplied as a high-purity reagent for in vitro studies.
  • Selective Akt/PKB inhibition for pathway studies.
  • Binds the pleckstrin homology (PH) domain.
  • Reduces phosphorylated Akt and cell proliferation in assays.
  • High purity (≈99.6%) suitable for biochemical experiments.
  • Small package sizes available for screening and pilot studies.
  • Molecular weight 337.29 g/mol; formula C13H15N5O6.