Learn More
Thermo Scientific API-II METAL NEEDLE KIT
Catalog No. NC9795041 Shop All Thermo Scientific Products
Encompass
API-2 metal needle kit, used in electrospray mode with metal needle option
Catalog No. NC9795041 Supplier Thermo Scientific Supplier No. OPTON53003
API-2 metal needle kit, used in electrospray mode with metal needle option