Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Waters MS Calibration Solution, Atmospheric Pressure Ionization, 15 mL, 18 Months Shelf Life

Catalog No. NC9161881 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

This standard is used for Mass Spec calibration assessment, adjusting mass calibration scales and relative ion intensities to known entities. Essential for all mass spectrometers, it addresses how subtle changes in electronics, surface cleanliness, and lab conditions affect instrument performance. While calibration is infrequent for nominal-mass instruments, response checks are vital, with close monitoring required for high accuracy.

  • Composition: NaCsI: Sodium Iodide 2μg/μL and Cesium Iodide 50ng/μL in 50/50 IPA/Water.
  • Volume: 15 mL.
  • Container: Durable, leak-proof, easy to carry, high capacity, wide-mouthed, and graduated bottle.
  • Shelf life: 18 Months.
  • Storage condition: Room temperature.
  • Shipping condition: Ambient.

Catalog No. NC9161881 Supplier Waters Corp Supplier No. 700001593
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This standard is used for Mass Spec calibration assessment, adjusting mass calibration scales and relative ion intensities to known entities. Essential for all mass spectrometers, it addresses how subtle changes in electronics, surface cleanliness, and lab conditions affect instrument performance. While calibration is infrequent for nominal-mass instruments, response checks are vital, with close monitoring required for high accuracy.

  • Composition: NaCsI: Sodium Iodide 2μg/μL and Cesium Iodide 50ng/μL in 50/50 IPA/Water.
  • Volume: 15 mL.
  • Container: Durable, leak-proof, easy to carry, high capacity, wide-mouthed, and graduated bottle.
  • Shelf life: 18 Months.
  • Storage condition: Room temperature.
  • Shipping condition: Ambient.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.