Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Waters Corporation Atmospheric Pressure Ionization Test Solution, Reversed Phase, MS Detection

Catalog No. 50818839 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

This standards kit is designed for system verification of various mass spectrometry instruments, including ZMD, QTofI/II/APIUS/Micro, PlatformII, QuattroII, and LCT.

  • Separation mode: Reversed phase
  • Detection method: MS
  • Formulation: Liquid, solid
  • Storage condition: Room temperature
  • System type: MS
  • Shelf life: 18 months
  • Shipping condition: Ambient
  • Type: Reference
  • Application: System performance
  • Product type: Standards
  • Units per package: 1 pk
  • Contains hazardous material requiring special freight handling

Catalog No. 50-818-839 Supplier Waters Corp Supplier No. 700000889
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This standards kit is designed for system verification of various mass spectrometry instruments, including ZMD, QTofI/II/APIUS/Micro, PlatformII, QuattroII, and LCT.

  • Separation mode: Reversed phase
  • Detection method: MS
  • Formulation: Liquid, solid
  • Storage condition: Room temperature
  • System type: MS
  • Shelf life: 18 months
  • Shipping condition: Ambient
  • Type: Reference
  • Application: System performance
  • Product type: Standards
  • Units per package: 1 pk
  • Contains hazardous material requiring special freight handling

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.