Learn More
Agilent Technologies API-TOF Reference Mass Solution Kit, 6 ampoules, 2.2 mL per ampoule, contains 2.5 mM Hexakis(1H, 1H, 3H-tetrafluoropropoxy)phosphazine, 5.0 mM Purine, 100.0 mM Ammonium Trifluoroacetate
Catalog No. NC2748341
Encompass
API-TOF Reference Mass Solution Kit, 6 ampoules, 2.2 mL per ampoule, contains 2.5 mM Hexakis(1H, 1H, 3H-tetrafluoropropoxy)phosphazine, 5.0 mM Purine, 100.0 mM Ammonium Trifluoroacetate
Catalog No. NC2748341 Supplier Agilent Technologies Supplier No. G196985001
API-TOF Reference Mass Solution Kit, 6 ampoules, 2.2 mL per ampoule, contains 2.5 mM Hexakis(1H, 1H, 3H-tetrafluoropropoxy)phosphazine, 5.0 mM Purine, 100.0 mM Ammonium Trifluoroacetate