Learn More
Medchemexpress LLC Api137 | 1428789-03-0 | 2291.71 | 5 MG
SDP
Catalog No. 5000398031
Encompass_Preferred
Api137 is an antimicrobial peptide that interferes with bacterial growth by inhibiting translation. It inhibits protein synthesis by trapping release factors on the 70S ribosome following hydrolysis of the nascent polypeptide chain.
  • Acts as an antimicrobial peptide
  • Interferes with bacterial growth by inhibiting translation
  • Inhibits protein synthesis by trapping release factors on the 70S ribosome
  • Follows hydrolysis of the nascent polypeptide chain
Catalog No. 50-003-98031 Supplier Medchemexpress LLC Supplier No. HYP5723A5MG
Api137 is an antimicrobial peptide that interferes with bacterial growth by inhibiting translation. It inhibits protein synthesis by trapping release factors on the 70S ribosome following hydrolysis of the nascent polypeptide chain.
  • Acts as an antimicrobial peptide
  • Interferes with bacterial growth by inhibiting translation
  • Inhibits protein synthesis by trapping release factors on the 70S ribosome
  • Follows hydrolysis of the nascent polypeptide chain