Learn More
Enzo Life Sciences Apicidin (5mg). CAS: 183506-66-3
SDP
Supplier:  Enzo Life Sciences BMLGR3400005
Encompass_Preferred
Potent inhibitor of histone deacetylase (HDAC). Inhibits proliferation. Induces cell cycle arrest at the G1 phase. At 100nM it induces a long lasting hyperacetylation of histone H4 while that induced by trichostatin is transient. Stimulates apoptosis. Apoptosis is induced via induction of Fas/Fas ligand. Displays potent antiangiogenic effects and dramatically decreases HIF-1α protein levels and transcriptional activity in human and mouse tumor cell lines. Antiprotozoal. Alternative name: cyclo-L-(2-Amino-8-oxodecanoyl)-L-(N-methoxy-tryptophan)-L-isoleucyl-D-pipecolinyl, Cyclo(N-O-methyl-L-tryptophanyl-L-isoleucinyl-D-pipecolinyl-L-2-amino-8-oxodecanoyl). Purity: ≥99% (HPLC). Solubility: Soluble in DMSO or 100% ethanol. Long Term Storage: -20°C.
Catalog No. 50-201-0995