Learn More
Abcam Apicidin, HDAC inhibitor, 1MG
Catalog No. 502706358
Encompass_Preferred
MW 623.8 Da, Purity >95%. Potent histone deacetylase inhibitor (IC₅₀ = 0.7 nM). Inhibits cancer cell proliferation via induction of p21WAF1/Cip1 and gelsolin. Active in vitro and in vivo. Cell-permeable.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6358 Supplier Abcam Supplier No. AB1420601MG
MW 623.8 Da, Purity >95%. Potent histone deacetylase inhibitor (IC₅₀ = 0.7 nM). Inhibits cancer cell proliferation via induction of p21WAF1/Cip1 and gelsolin. Active in vitro and in vivo. Cell-permeable.
The product is subject to the following: Abcam Restricted Use Statement