Learn More
Electron Microscopy Sciences Apiezon High Vacuum AP100 Grease 50 GR
SDP
Catalog No. 501902646
Encompass_Preferred
Apiezon AP100 Grease is a silicone-free vacuum grease and lubricant. Containing PTFE, AP100 exhibits extremely high levels of lubricity and has been shown to provide eight times the level of lubrication offered by standard, petroleum-based lubricating greases.
Apiezon AP100 is designed for use at ambient temperatures (generally between 10 to 30 degree C / 50 to 86 degree F).
The unique properties of Apiezon AP100 lubricating vacuum grease mean it is frequently used to protect stepping motors and gearboxes from corrosion and abrasion, particularly when they are subjected to high load conditions. AP100 contains "PTFE" which confers superior anti-seize properties providing long lasting lubrication and ensuring smooth operation of stop cocks and taps. It demonstrates an extremely low vapour pressure of 7 x 10-11 Torr at 20 degree C.
Catalog No. 50-190-2646 Supplier Electron Microscopy Sciences Supplier No. 60709
Apiezon AP100 Grease is a silicone-free vacuum grease and lubricant. Containing PTFE, AP100 exhibits extremely high levels of lubricity and has been shown to provide eight times the level of lubrication offered by standard, petroleum-based lubricating greases.
Apiezon AP100 is designed for use at ambient temperatures (generally between 10 to 30 degree C / 50 to 86 degree F).
The unique properties of Apiezon AP100 lubricating vacuum grease mean it is frequently used to protect stepping motors and gearboxes from corrosion and abrasion, particularly when they are subjected to high load conditions. AP100 contains "PTFE" which confers superior anti-seize properties providing long lasting lubrication and ensuring smooth operation of stop cocks and taps. It demonstrates an extremely low vapour pressure of 7 x 10-11 Torr at 20 degree C.