Learn More
Electron Microscopy Sciences Apiezon High Vacuum Grease AP101 50 GR
SDP
Catalog No. 50365103
Encompass_Preferred
Type 101 is an excellent general purpose, hydrocarbon grease, which is intended for a variety of industrial and scientific applications. The AP 101 contains "PTFE" which confers superior anti-seize properties providing long lasting lubrication and ensuring smooth operation of stop cocks and taps. The value of AP 101 anti-seize properties are not limited to laboratory.
Catalog No. 50-365-103 Supplier Electron Microscopy Sciences Supplier No. 6070601
Type 101 is an excellent general purpose, hydrocarbon grease, which is intended for a variety of industrial and scientific applications. The AP 101 contains "PTFE" which confers superior anti-seize properties providing long lasting lubrication and ensuring smooth operation of stop cocks and taps. The value of AP 101 anti-seize properties are not limited to laboratory.