Learn More
Electron Microscopy Sciences Apiezon High Vacuum Grease, Type H 25 GR
SDP
Supplier:  Electron Microscopy Sciences 60703
Encompass_Preferred
Type H grease is designed for high temperatures (-10 to +240 degree C) in a wide range of applications in both science and industry. With good "friction", grease H is ideal for use with laboratory glassware, but combined with properties of high thermal conductivity, it is the perfect choice for the electronics and space industries where heat sink media require adhesion.
Grease H is a filled hydrocarbon which exhibits excellent thermal conductivity, and strong absorption properties. The capability to absorb greasy or chemical impurities on metal and glass surfaces is a value "tool" required by electronic industries.
Catalog No. 50-365-261