Learn More
Electron Microscopy Sciences Apiezon High Vacuum Grease Type M 100 GR
SDP
Supplier:  Electron Microscopy Sciences 6070001
Encompass_Preferred
Similar to grease type L, but it contains more wax. Grease M has an estimated vapor pressure @ 20 degree C of 2x10-9 torr, higher than Type L. Type M is recommended for applications where a grease of moderately low vapor pressure is required; such as for sealing ground joints on the backing side of oil diffusion pumps.
Catalog No. 50-193-1177