Learn More
Electron Microscopy Sciences Apiezon High Vacuum Grease, Type N 25 GR
SDP
Supplier:  Electron Microscopy Sciences 60701
Encompass_Preferred
Similar to Type L and Type M, but Type N grease has a special additive which gives it a tenacious, rubbery consistency and provides an extra cushioning effect which absorbs vibrations in equipment, making N grease invaluable in fragile glass to glass joints, like burette taps, which continually risk fracture. Grease N is widely recognized and recommended as the cryogenic vacuum grease of choice.
Catalog No. 50-189-8973