Learn More
Electron Microscopy Sciences Invoil Diffusion Pump Oil BW 500 ML
SDP
Catalog No. 50996272
Encompass_Preferred
Consists of esters or specially selected hydrocarbon fractions. They were developed for vapor-diffusion pump fluids. They are high purity, very low pressure, and very stable. Pressures lower than 10^-7 torr obtainable. No need for cold trap. Good pumping speed and no corrosive deposits or vapors. Water-white oil with a higher pumping speed than B. High purity suitable for evacuation of mass spectrometers.
Catalog No. 50-996-272 Supplier Electron Microscopy Sciences Supplier No. 60722
Consists of esters or specially selected hydrocarbon fractions. They were developed for vapor-diffusion pump fluids. They are high purity, very low pressure, and very stable. Pressures lower than 10^-7 torr obtainable. No need for cold trap. Good pumping speed and no corrosive deposits or vapors. Water-white oil with a higher pumping speed than B. High purity suitable for evacuation of mass spectrometers.