Learn More
Electron Microscopy Sciences Apiezon High Vacuum PFPE 501 Grease 100 GR
SDP
Supplier:  Electron Microscopy Sciences 60708
Encompass_Preferred
PFPE 501 is a chemically inert, high temperature, and extreme pressure lubricant. With an upper operating temperature of 250 degree C and vapour pressure of 1.3 x 10-12 at 25 degree C, it is robust, versatile and can be used for sealing and lubricating under the most extreme operating conditions. This includes environments where aggressive chemicals and strong oxidising agents are regularly used. It is PFPE based, used in oxygen rich environments, ultra high vacuum, and has a wide temperature range. It extends equipment life, reduces equipment downtime, lowers service costs, and has expert technical support.
Catalog No. 50-190-2645