Learn More
M&I Materials Apiezon™ Sealing Compound Q
Catalog No. 1463820
Change view
Click to view available options
Product Type:
Sealing Compound Q
Catalog No. Product Type
14-638-20 Sealing Compound Q
1 options
Catalog No. 14-638-20 Supplier M&I Materials Supplier No. FAP/W/Q01000
Add to Cart
Add to Cart
Effective temporary vacuum sealant that is used as a short term solution in situations which do not allow for the immediate dismantling of a system for repair.
Putty-like sealing compound easily molds to any shape. For medium vacuum systems. Vapor pressure 10-2 Pa (10-4 torr) at 20°C (68°F). working temperature range -10°C to +30°C.
Order Info
Can of 1kg.
Specifications
Product Type Sealing Compound Q
For Use With (Equipment) Medium vacuum system
Pressure 104 torr
Quantity 5 Pk.
Temperature (English) 14°F to 86°F
Temperature (Metric) -10°C to +30°C