Learn More
Electron Microscopy Sciences Apiezon Vacuum Sealing Wax Q Vacuum Sealing Compound 1 KG
SDP
Catalog No. 501902650
Encompass_Preferred
Q Compound is an effective temporary vacuum sealant that is used as a short term solution in situations which do not allow for the immediate dismantling of a system for repair.
Use: Vacuum Sealing Compound
Catalog No. 50-190-2650 Supplier Electron Microscopy Sciences Supplier No. 60713
Q Compound is an effective temporary vacuum sealant that is used as a short term solution in situations which do not allow for the immediate dismantling of a system for repair.
Use: Vacuum Sealing Compound