Learn More
Electron Microscopy Sciences Apiezon Vacuum Sealing Wax W100 Medium Hardness Vacuum Sealant 250 GR
SDP
Catalog No. 501902648
Encompass_Preferred
Easy to use Apiezon waxes can be used as permanent or semi-permanent vacuum sealants or mounting media. The waxes have a high purity combined with powerful gettering and clean meltdown properties. An excellent waterproof media that is easy to use and to remove. 10 year shelf life. Wax W100 is a medium hardness vacuum sealant and mounting wax.
Use: Medium Hardness Vacuum Sealant / Mounting Wax
Catalog No. 50-190-2648 Supplier Electron Microscopy Sciences Supplier No. 60711
Easy to use Apiezon waxes can be used as permanent or semi-permanent vacuum sealants or mounting media. The waxes have a high purity combined with powerful gettering and clean meltdown properties. An excellent waterproof media that is easy to use and to remove. 10 year shelf life. Wax W100 is a medium hardness vacuum sealant and mounting wax.
Use: Medium Hardness Vacuum Sealant / Mounting Wax