Learn More
Electron Microscopy Sciences Apiezon Vacuum Sealing Wax W40 Soft Vacuum Sealant 250 GR
SDP
Supplier:  Electron Microscopy Sciences 60712
Encompass_Preferred
Easy to use Apiezon waxes can be used as permanent or semi-permanent vacuum sealants or mounting media. The waxes have a high purity combined with powerful gettering and clean meltdown properties. An excellent waterproof media that is easy to use and to remove. 10 year shelf life. Wax W40 is a soft vacuum sealant and mounting wax.
Use: Soft Vacuum Sealant / Mounting Wax
Catalog No. 50-190-2649