Learn More
Sigma Aldrich Fine Chemicals Biosciences ApiezonR grease1EA
Catalog No. 502904991
Encompass
for low vapor pressure sealing and lubrication in high vacuum applications.
Catalog No. 50-290-4991 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z2735621EA
for low vapor pressure sealing and lubrication in high vacuum applications.