Learn More
Sigma Aldrich Fine Chemicals Biosciences ApiezonR grease1EA
Supplier: Sigma Aldrich Fine Chemicals Biosciences Z2735891EA
for low vapor pressure sealing and lubrication in high vacuum applications.