Learn More
Medchemexpress LLC HY-N1201 100mg , Apigenin CAS:520-36-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN1201100MG
Encompass_Preferred
HY-N1201 100mg Apigenin CAS: 520-36-5 Apigenin (4',5,7-Trihydroxyflavone) is a competitive CYP2C9 inhibitor with a Ki of 2 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-1181