Learn More
AdipoGen Apigenin (25 mg)
SDP
Supplier:  AdipoGen CDXA0438M025
Encompass_Preferred
Chemical. CAS: 520-36-5. Formula: C15H10O5. MW: 270.24. Isolated from plant source. Apigenin is an antioxidant plant flavonoid with anti-inflammatory, angiogenic, neuroprotective and anticancer properties. It may induce apoptosis and inhibit proliferation of tumor cells by arresting the cell cycle at the G2/M phase. Inhibits tumor angiogenesis through decreasing HIF-1alpha and VEGF expression. Also known to be a MAP kinase inhibitor and a selectively inhibitor of casein kinase 2 (CK2). It is a potent inhibitor of the synthesis of the inflammatory mediators nitric oxide and prostaglandin E2, reducing inducible nitric oxide synthase (iNOS) and cyclooxygenase-2 (COX-2) expression.
Catalog No. 50-205-4272