Learn More
Medchemexpress LLC Apigenin (4',5,7-Trihydroxyflavone) | 520-36-5 | 270.24 | 5 G
SDP
Catalog No. 5000415741
Encompass_Preferred
Apigenin (4',5,7-Trihydroxyflavone) is a competitive CYP2C9 inhibitor with a Ki of 2 μM. This natural flavonoid possesses broad biological properties including antioxidative, anti-inflammatory, anticancer, and neuroprotective effects.
  • Competitive CYP2C9 inhibitor
  • Exhibits antioxidative effects
  • Exhibits anti-inflammatory effects
  • Exhibits anticancer effects
  • Exhibits neuroprotective effects
  • Inhibits cell proliferation
  • Alleviates Adriamycin (ADR)-induced myocardial injury
Catalog No. 50-004-15741 Supplier Medchemexpress LLC Supplier No. HYN12015G
Apigenin (4',5,7-Trihydroxyflavone) is a competitive CYP2C9 inhibitor with a Ki of 2 μM. This natural flavonoid possesses broad biological properties including antioxidative, anti-inflammatory, anticancer, and neuroprotective effects.
  • Competitive CYP2C9 inhibitor
  • Exhibits antioxidative effects
  • Exhibits anti-inflammatory effects
  • Exhibits anticancer effects
  • Exhibits neuroprotective effects
  • Inhibits cell proliferation
  • Alleviates Adriamycin (ADR)-induced myocardial injury