Learn More
Medchemexpress LLC Apigenin (4',5,7-Trihydroxyflavone) | 520-36-5 | 99.2% | 50 MG
SDP
Catalog No. 5000366190
Encompass_Preferred
Apigenin (4',5,7-Trihydroxyflavone) is a competitive CYP2C9 inhibitor and inhibits cell proliferation. It suppresses the clone formation of SGC-7901 cells in a dose- and time-dependent manner.
  • Competitive CYP2C9 inhibitor
  • Inhibits cell proliferation
  • Suppresses SGC-7901 cell clone formation
Catalog No. 50-003-66190 Supplier Medchemexpress LLC Supplier No. 50MGHYN1201
Apigenin (4',5,7-Trihydroxyflavone) is a competitive CYP2C9 inhibitor and inhibits cell proliferation. It suppresses the clone formation of SGC-7901 cells in a dose- and time-dependent manner.
  • Competitive CYP2C9 inhibitor
  • Inhibits cell proliferation
  • Suppresses SGC-7901 cell clone formation