Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Apigenin (4',5,7-Trihydroxyflavone) | 520-36-5 | HY-N1201 | 98.9% | 270.24 | 500 MG
SDP

Catalog No. 5000365879
Encompass_Preferred

Apigenin (4',5,7-Trihydroxyflavone) is identified as a competitive CYP2C9 inhibitor with a Ki of 2 μM. It demonstrates the ability to inhibit cell proliferation, with growth inhibition rates of 38%, 71%, and 99% at concentrations of 20, 40, and 80 μM respectively. The compound also suppresses clone formation in SGC-7901 cells in a dose- and time-dependent manner.

  • Competitive inhibitor of Cytochrome P450 2C9 (CYP2C9).
  • Exhibits cell proliferation inhibitory activity.
  • Suppresses clone formation in specific cell lines.

Catalog No. 50-003-65879 Supplier Medchemexpress LLC Supplier No. 500MGHYN1201
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Apigenin (4',5,7-Trihydroxyflavone) is identified as a competitive CYP2C9 inhibitor with a Ki of 2 μM. It demonstrates the ability to inhibit cell proliferation, with growth inhibition rates of 38%, 71%, and 99% at concentrations of 20, 40, and 80 μM respectively. The compound also suppresses clone formation in SGC-7901 cells in a dose- and time-dependent manner.

  • Competitive inhibitor of Cytochrome P450 2C9 (CYP2C9).
  • Exhibits cell proliferation inhibitory activity.
  • Suppresses clone formation in specific cell lines.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.