Learn More
Medchemexpress LLC Apigenin | 520-36-5 | 1 ML
SDP
Catalog No. 5000415145
Encompass_Preferred
Apigenin, also known as 4',5,7-Trihydroxyflavone, functions as a competitive inhibitor of CYP2C9 with a Ki of 2 μM. This compound has been observed to inhibit cell proliferation and suppress clone formation in various cell lines, making it suitable for research applications exploring its biological activity.
  • Competitive CYP2C9 inhibitor
  • Inhibits cell proliferation
  • Suppresses clone formation of cells
Catalog No. 50-004-15145 Supplier Medchemexpress LLC Supplier No. HYN120110MM/1ML
Apigenin, also known as 4',5,7-Trihydroxyflavone, functions as a competitive inhibitor of CYP2C9 with a Ki of 2 μM. This compound has been observed to inhibit cell proliferation and suppress clone formation in various cell lines, making it suitable for research applications exploring its biological activity.
  • Competitive CYP2C9 inhibitor
  • Inhibits cell proliferation
  • Suppresses clone formation of cells