Learn More
Medchemexpress LLC Apigenin | 520-36-5 | 270.24 | 5 MG
SDP
Catalog No. 5000416330
Encompass_Preferred
Apigenin (4',5,7-Trihydroxyflavone) is a competitive CYP2C9 inhibitor with a Ki of 2 μM. It inhibits cell proliferation in SGC-7901 cells in a dose- and time-dependent manner and possesses a broad spectrum of biological properties including antioxidative, anti-inflammatory, anticancer, and neuroprotective effects.
  • Inhibits cell proliferation
  • Suppresses clone formation of SGC-7901 cells
  • Alleviates adriamycin-induced myocardial injury
  • Reduces serum aspartate amino transferase release
  • Reduces serum lactate dehydrogenase release
  • Reduces serum creatine kinase contents
Catalog No. 50-004-16330 Supplier Medchemexpress LLC Supplier No. HYN12015MG
Apigenin (4',5,7-Trihydroxyflavone) is a competitive CYP2C9 inhibitor with a Ki of 2 μM. It inhibits cell proliferation in SGC-7901 cells in a dose- and time-dependent manner and possesses a broad spectrum of biological properties including antioxidative, anti-inflammatory, anticancer, and neuroprotective effects.
  • Inhibits cell proliferation
  • Suppresses clone formation of SGC-7901 cells
  • Alleviates adriamycin-induced myocardial injury
  • Reduces serum aspartate amino transferase release
  • Reduces serum lactate dehydrogenase release
  • Reduces serum creatine kinase contents