Learn More
Medchemexpress LLC Apigenin-7-diglucuronide | 74696-01-8 | 1 MG
SDP
Catalog No. 5000419409
Encompass_Preferred
Apigenin-7-diglucuronide is a flavonoid glycoside found in various medicinal plants. It exhibits anti-inflammatory and antioxidant activities, and has been shown to protect retinas against bright light-induced photoreceptor degeneration by inhibiting retinal oxidative stress and inflammation.
  • Flavonoid glycoside
  • Found in various medicinal plants
  • Exhibits anti-inflammatory and antioxidant activities
  • Protects retinas against bright light-induced photoreceptor degeneration
  • Inhibits retinal oxidative stress and inflammation
  • Intended for research use only
Catalog No. 50-004-19409 Supplier Medchemexpress LLC Supplier No. HYN72701MG
Apigenin-7-diglucuronide is a flavonoid glycoside found in various medicinal plants. It exhibits anti-inflammatory and antioxidant activities, and has been shown to protect retinas against bright light-induced photoreceptor degeneration by inhibiting retinal oxidative stress and inflammation.
  • Flavonoid glycoside
  • Found in various medicinal plants
  • Exhibits anti-inflammatory and antioxidant activities
  • Protects retinas against bright light-induced photoreceptor degeneration
  • Inhibits retinal oxidative stress and inflammation
  • Intended for research use only