Learn More
TARGETMOL CHEMICALS INC APIGENIN 7-GLUCOSIDE 25MG
SDP
Catalog No. 502579836
Encompass_Preferred
Also available in 5 mg 10 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. 1. Apigenin 7-glucoside (Cosmosiin) is a anti-HIV agent. 2. Cosmosiin may be beneficial for diabetic complications through their enhanced adiponectin secretion tyrosine phosphorylation of insulin receptor-(beta) and GLUT4 translocation. purity: 99%
Catalog No. 50-257-9836 Supplier TARGETMOL CHEMICALS INC Supplier No. T4S029525MG
Also available in 5 mg 10 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. 1. Apigenin 7-glucoside (Cosmosiin) is a anti-HIV agent. 2. Cosmosiin may be beneficial for diabetic complications through their enhanced adiponectin secretion tyrosine phosphorylation of insulin receptor-(beta) and GLUT4 translocation. purity: 99%