Learn More
Cayman Chemical Apigenin 7-glucoside, Purity: ≥98%, CAS: 578-74-5, 25 mg
SDP
Catalog No. 502935224
Encompass_Preferred
A flavonoid with anti-inflammatory and anxiolytic activities; inhibits LPS-induced nitric oxide production in RAW 264.7 cells from 0.16-10 μM; reduces pulmonary edema and lung inflammation in a mouse model of LPS-induced acute lung injury at 10 mg/kg; increases the number of entries and the time spent in the open arms of the elevated plus maze in rats
Catalog No. 50-293-5224 Supplier Cayman Chemical Supplier No. 2681325 MG
A flavonoid with anti-inflammatory and anxiolytic activities; inhibits LPS-induced nitric oxide production in RAW 264.7 cells from 0.16-10 μM; reduces pulmonary edema and lung inflammation in a mouse model of LPS-induced acute lung injury at 10 mg/kg; increases the number of entries and the time spent in the open arms of the elevated plus maze in rats