Learn More
Medchemexpress LLC Apigenin 7-glucoside | 578-74-5 | 432.38 | 25 MG
SDP
Catalog No. 5000390185
Encompass_Preferred
Apigenin-7-glucoside (Apigenin-7-O-β-D-glucopyranoside) demonstrates notable anti-proliferative and antioxidant properties, and is effective in scavenging reactive oxygen species (ROS). It is intended for research use only and is not sold to patients.
  • Exhibits significant anti-proliferative activity against B16F10 melanoma cells.
  • Provokes an increase of subG0/G1, S, and G2/M phase cell proportion with a significant decrease of cell proportion in G0/G1 phases.
  • Enhances melanogenesis synthesis and tyrosinase activity of B16F10 melanoma cells.
  • Specifically induces the differentiation of CD34+ cells towards the erythroid lineage and inhibits myeloid differentiation.
  • Possesses strong antioxidant activity against reactive oxygen species (ROS) in vitro in a concentration-dependent manner.
Catalog No. 50-003-90185 Supplier Medchemexpress LLC Supplier No. 25MGHYN0578
Apigenin-7-glucoside (Apigenin-7-O-β-D-glucopyranoside) demonstrates notable anti-proliferative and antioxidant properties, and is effective in scavenging reactive oxygen species (ROS). It is intended for research use only and is not sold to patients.
  • Exhibits significant anti-proliferative activity against B16F10 melanoma cells.
  • Provokes an increase of subG0/G1, S, and G2/M phase cell proportion with a significant decrease of cell proportion in G0/G1 phases.
  • Enhances melanogenesis synthesis and tyrosinase activity of B16F10 melanoma cells.
  • Specifically induces the differentiation of CD34+ cells towards the erythroid lineage and inhibits myeloid differentiation.
  • Possesses strong antioxidant activity against reactive oxygen species (ROS) in vitro in a concentration-dependent manner.