Learn More
Medchemexpress LLC Apigenin 7-glucoside | 578-74-5 | MFCD00016787 | 432.38 | 50 MG
SDP
Catalog No. 5000388560
Encompass_Preferred
Apigenin-7-glucoside (Apigenin-7-O-β-D-glucopyranoside) exhibits significant anti-proliferative and antioxidant activity and scavenges reactive oxygen species (ROS). It is for research use only and not sold to patients. It is available as a solid, light yellow to yellow in color.
  • Exhibits significant anti-proliferative and antioxidant activity
  • Scavenges reactive oxygen species (ROS)
  • Induces differentiation of CD34+ cells towards erythroid lineage
  • Inhibits myeloid differentiation
  • Available as a solid, light yellow to yellow in color
Catalog No. 50-003-88560 Supplier Medchemexpress LLC Supplier No. HYN057850MG
Apigenin-7-glucoside (Apigenin-7-O-β-D-glucopyranoside) exhibits significant anti-proliferative and antioxidant activity and scavenges reactive oxygen species (ROS). It is for research use only and not sold to patients. It is available as a solid, light yellow to yellow in color.
  • Exhibits significant anti-proliferative and antioxidant activity
  • Scavenges reactive oxygen species (ROS)
  • Induces differentiation of CD34+ cells towards erythroid lineage
  • Inhibits myeloid differentiation
  • Available as a solid, light yellow to yellow in color