Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Apigenin 7-glucoside, ≥97.0% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111014732 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
5 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-4732 5 mg
11-101-4733 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-101-4732 Supplier MilliporeSigma Supelco Supplier No. 446925MGF
Add to Cart
Add to Cart

Apigenin 7-glucoside is a flavone belonging to the class of polyphenols and is commonly found in Thymus species. It shows a broad spectrum of pharmacological properties such as antispasmodic, bactericidal, expectorant, antihelmintic, adstringent, antitussive and carminative agent.

Food and Beverages
TRUSTED_SUSTAINABILITY

Specifications

Packaging Bottomless Glass Bottle with Inserted Fused Cone
CAS 578-74-5
Physical Form Neat
Linear Formula C21H20O10
MDL Number MFCD00016787
Synonym 4 ′,5,7-Trihydroxyflavone 7-glucoside; Apigetrin; Cosmosiin
Type Analytical Standard
Quantity 5 mg
Formula Weight 432.38
Percent Purity ≥97.0% (HPLC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.