Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Apigenin-7-glucuronide | 29741-09-1 | 446.36 | 1 ML
SDP

Catalog No. 5000414529
Encompass_Preferred

Apigenin-7-glucuronide is a chemical compound that acts as an inhibitor of Matrix Metalloproteinases (MMP) activities, with specific IC50 values for MMP-3, MMP-8, MMP-9, and MMP-13. This product is intended for research use only.

  • Inhibits Matrix Metalloproteinases (MMP) activities
  • Purity of 99.74%
  • Available as a solid
  • Soluble in DMSO for in vitro studies
  • Specific storage conditions for powder and in solvent

Catalog No. 50-004-14529 Supplier Medchemexpress LLC Supplier No. HYN145410MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Apigenin-7-glucuronide is a chemical compound that acts as an inhibitor of Matrix Metalloproteinases (MMP) activities, with specific IC50 values for MMP-3, MMP-8, MMP-9, and MMP-13. This product is intended for research use only.

  • Inhibits Matrix Metalloproteinases (MMP) activities
  • Purity of 99.74%
  • Available as a solid
  • Soluble in DMSO for in vitro studies
  • Specific storage conditions for powder and in solvent

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.