Learn More
TARGETMOL CHEMICALS INC APIGENIN-7-GLUCURONIDE 5MG
SDP
Catalog No. 502576643
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Apigenin-7-glucuronide (Apigenin 7-O-glucuronide) possesses multiple activities including antioxidant anti-inflammatory and aldose reductase inhibitory activities it can inhibit MMP activities (IC50s 12.87 22.39 17.52 0.27 uM for MMP-3 MMP-8 MMP-9 MMP-13 respectively). purity: 99%
Catalog No. 50-257-6643 Supplier TARGETMOL CHEMICALS INC Supplier No. T5S10265MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Apigenin-7-glucuronide (Apigenin 7-O-glucuronide) possesses multiple activities including antioxidant anti-inflammatory and aldose reductase inhibitory activities it can inhibit MMP activities (IC50s 12.87 22.39 17.52 0.27 uM for MMP-3 MMP-8 MMP-9 MMP-13 respectively). purity: 99%