Learn More
Medchemexpress LLC HY-N1454 5mg Medchemexpress, Apigenin-7-glucuronide CAS:29741-09-1 Purity:>98%
SDP
Catalog No. 501933837
Encompass_Preferred
Medchemexpress, HY-N1454 5mg Apigenin-7-glucuronide CAS:29741-09-1 Apigenin-7-glucuronide could inhibit Matrix Metalloproteinases (MMP) activities, with IC 50 s of 12.87, 22.39, 17.52, 0.27 μM for MMP-3, MMP-8, MMP-9, MMP-13, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3837 Supplier Medchemexpress LLC Supplier No. HYN14545MG
Medchemexpress, HY-N1454 5mg Apigenin-7-glucuronide CAS:29741-09-1 Apigenin-7-glucuronide could inhibit Matrix Metalloproteinases (MMP) activities, with IC 50 s of 12.87, 22.39, 17.52, 0.27 μM for MMP-3, MMP-8, MMP-9, MMP-13, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.