Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Apigenin-7-O-(2G-rhamnosyl)gentiobioside | 174284-20-9 | 1 MG
SDP

Catalog No. 5000421411
Encompass_Preferred

Apigenin-7-O-(2G-rhamnosyl)gentiobioside is a flavone glycoside found in *Lonicera gracilipes* var. *glandulosa*. It has a molecular weight of 740.66 and a molecular formula of C33H40O19. This product is intended for research use only.

  • Solid appearance
  • Store at 4°C, protected from light
  • Soluble in DMSO at 50 mg/mL

Catalog No. 50-004-21411 Supplier Medchemexpress LLC Supplier No. HYN21531MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Apigenin-7-O-(2G-rhamnosyl)gentiobioside is a flavone glycoside found in *Lonicera gracilipes* var. *glandulosa*. It has a molecular weight of 740.66 and a molecular formula of C33H40O19. This product is intended for research use only.

  • Solid appearance
  • Store at 4°C, protected from light
  • Soluble in DMSO at 50 mg/mL

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.