Learn More
TARGETMOL CHEMICALS INC Apigenin-7-O-2G-rhamnosylgenti
SDP
Catalog No. 502879796
Encompass_Preferred
Also available in 1 mL, 1 mg, 10 mg, 25 mg, 50 mg and bulk. Please contact Fisher for quotes. Apigenin-7-O-(2G-rhamnosyl)gentiobioside is a flavonoid glycoside compound extracted from [Lonicera gracilipes var. glandulosa]. Purity 99.71%
Catalog No. 50-287-9796 Supplier TARGETMOL CHEMICALS INC Supplier No. T5S15265MG
Also available in 1 mL, 1 mg, 10 mg, 25 mg, 50 mg and bulk. Please contact Fisher for quotes. Apigenin-7-O-(2G-rhamnosyl)gentiobioside is a flavonoid glycoside compound extracted from [Lonicera gracilipes var. glandulosa]. Purity 99.71%