Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Apigenin-7-O-β-D-glucopyranoside | 578-74-5 | MFCD00016787 | 99.0% | 432.38 | C21H20O10 | 10 MG
SDP

Catalog No. 5000296978
Encompass_Preferred

Apigenin 7-glucoside (Apigenin-7-O-β-D-glucopyranoside) is a flavonoid glycoside used as an analytical standard and research reagent, exhibiting antioxidant and anti-proliferative activity and commonly employed in biochemical and analytical assays.

  • Natural product flavonoid glycoside.
  • Exhibits antioxidant and anti-proliferative activity.
  • Molecular formula C21H20O10, molecular weight 432.38.
  • CAS number 578-74-5.
  • High purity (≈99%) suitable for analytical and in vitro research.
  • Storage: store at 4°C, protect from light; in solvent: -80°C (2 years) or -20°C (1 year).

Catalog No. 50-002-96978 Supplier Medchemexpress LLC Supplier No. HYN057810MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Apigenin 7-glucoside (Apigenin-7-O-β-D-glucopyranoside) is a flavonoid glycoside used as an analytical standard and research reagent, exhibiting antioxidant and anti-proliferative activity and commonly employed in biochemical and analytical assays.

  • Natural product flavonoid glycoside.
  • Exhibits antioxidant and anti-proliferative activity.
  • Molecular formula C21H20O10, molecular weight 432.38.
  • CAS number 578-74-5.
  • High purity (≈99%) suitable for analytical and in vitro research.
  • Storage: store at 4°C, protect from light; in solvent: -80°C (2 years) or -20°C (1 year).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.