Learn More
Medchemexpress LLC Apigenin 7-O-glucuronide | 29741-09-1 | MFCD23699539 | >95.0% | 10 MG
SDP
Catalog No. 5000296483
Encompass_Preferred
Apigenin 7-O-glucuronide is a flavonoid glucuronide supplied as an analytical standard for research and analytical applications. It has reported inhibitory activity against matrix metalloproteinases, with IC50s of 12.87 μM (MMP-3), 22.39 μM (MMP-8), 17.52 μM (MMP-9), and 0.27 μM (MMP-13). CAS: 29741-09-1; molecular weight: 446.36 g/mol.
  • Provided as a 10 mg analytical standard for research use only.
  • Reported inhibitory activity against multiple MMP enzymes with published IC50 values.
  • Chemical formula C21H18O11 and molecular weight 446.36 g/mol.
  • Commonly used in biochemical and analytical assays to characterize apigenin metabolites.
Catalog No. 50-002-96483 Supplier Medchemexpress LLC Supplier No. HYN145410MG
Apigenin 7-O-glucuronide is a flavonoid glucuronide supplied as an analytical standard for research and analytical applications. It has reported inhibitory activity against matrix metalloproteinases, with IC50s of 12.87 μM (MMP-3), 22.39 μM (MMP-8), 17.52 μM (MMP-9), and 0.27 μM (MMP-13). CAS: 29741-09-1; molecular weight: 446.36 g/mol.
  • Provided as a 10 mg analytical standard for research use only.
  • Reported inhibitory activity against multiple MMP enzymes with published IC50 values.
  • Chemical formula C21H18O11 and molecular weight 446.36 g/mol.
  • Commonly used in biochemical and analytical assays to characterize apigenin metabolites.