Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Apigenin 7-O-malonylglucoside | 86546-87-4 | 1 MG
SDP

Catalog No. 5000420891
Encompass_Preferred

Apigenin 7-O-malonylglucoside is a flavonoid found in chrysanthemum flowers. It is for research use only.

  • Purity >98%
  • Appearance: solid
  • Color: white to off-white
  • Molecular weight: 518.42
  • Chemical formula: C24H22O13
  • Storage: 4°C, protect from light
  • Storage in solvent: -80°C, protect from light
  • Soluble in DMSO

Catalog No. 50-004-20891 Supplier Medchemexpress LLC Supplier No. HYN24961MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Apigenin 7-O-malonylglucoside is a flavonoid found in chrysanthemum flowers. It is for research use only.

  • Purity >98%
  • Appearance: solid
  • Color: white to off-white
  • Molecular weight: 518.42
  • Chemical formula: C24H22O13
  • Storage: 4°C, protect from light
  • Storage in solvent: -80°C, protect from light
  • Soluble in DMSO

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.