Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Apigenin 7-O-malonylglucoside | 86546-87-4 | 5 MG
SDP

Catalog No. 5000421339
Encompass_Preferred

Apigenin 7-O-malonylglucoside is a compound found in chrysanthemum flowers. This product is intended for research use only and is not for human use or medical applications.

  • White to off-white solid appearance
  • Purity of 97.29%
  • Store at 4°C, protected from light
  • In solvent, store at -80°C for 6 months or -20°C for 1 month, protected from light
  • Soluble in DMSO at 50 mg/mL (96.45 mM) with ultrasonic required

Catalog No. 50-004-21339 Supplier Medchemexpress LLC Supplier No. HYN24965MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Apigenin 7-O-malonylglucoside is a compound found in chrysanthemum flowers. This product is intended for research use only and is not for human use or medical applications.

  • White to off-white solid appearance
  • Purity of 97.29%
  • Store at 4°C, protected from light
  • In solvent, store at -80°C for 6 months or -20°C for 1 month, protected from light
  • Soluble in DMSO at 50 mg/mL (96.45 mM) with ultrasonic required

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.